missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Antibody (SR1498), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | Thyroglobulin |
|---|---|
| Clone | SR1498 |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600884
|
Novus Biologicals
NBP3-21918-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678254
|
Novus Biologicals
NBP3-21918-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| Thyroglobulin | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AITD3TGN, TDH3, Tg, thyroglobulin | |
| A synthesized peptide derived from human Thyroglobulin (Uniprot #: P01266) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1498 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 7038 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title