missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Thyroglobulin |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 6E1 |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 0.1 - 0.2 μg/mL |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | AITD3TGN, TDH3, Tg, thyroglobulin |
| Host Species | Mouse |
| Immunogen | Human thyroid follicular cells |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?