missing translation for 'onlineSavingsMsg'
Learn More

Thyroglobulin Antibody (2H11 + 6E1) - Azide and BSA Free, Novus Biologicals™

Product Code. 18406181 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406181 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406181 Supplier Novus Biologicals Supplier No. NBP2345300.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thyroglobulin
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone 2H11 + 6E1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 to 1 μg/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin 0.5 to 1 μg/mL, CyTOF-ready
Formulation 10 mM PBS
Gene Alias AITD3TGN, TDH3, Tg, thyroglobulin
Host Species Mouse
Immunogen Human thyroid follicular cells
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 7038
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.