Learn More
THYN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558523
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KGLSGKRTKT ENSGEALAKV EDSNPQKTSA TKNCLKNLSS HWLMKSEPES RLEKGVDVKF SIEDLKAQPK QTTCWDGVRN YQARNFLRAM KLGEEA Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.
THYN1, also named as THY28, is a 28-32 kDa protein which located mainly in the nucleus. It appears to correlate with induction of apoptosis.Specifications
THYN1 | |
Polyclonal | |
Unconjugated | |
THYN1 | |
D730042P09Rik; HSPC144; MDS012; mThy28; My0054; MY105; Thy28; THY28KD; Thymocyte nuclear protein 1; thymocyte protein thy28; Thyn1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29087 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P016 | |
THYN1 | |
Recombinant protein corresponding to Human THYN1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only