missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THYN1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98501PE
This item is not returnable.
View return policy
Description
THYN1 Polyclonal antibody specifically detects THYN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| THYN1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PE | |
| HSPC144, MDS012, MGC12187, THY28KD, THY28MY105, thymocyte nuclear protein 1, Thymocyte protein Thy28 | |
| Produced in rabbits immunized with E. coli-derived Human THYN1 fragment. | |
| 0.1 mL | |
| Apoptosis | |
| 29087 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction