missing translation for 'onlineSavingsMsg'
Learn More

Thymosin beta 4 Antibody, Novus Biologicals™

Product Code. 18363618 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363618 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363618 Supplier Novus Biologicals Supplier No. H00007114B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Thymosin beta 4 Polyclonal antibody specifically detects Thymosin beta 4 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thymosin beta 4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.4)
Gene Accession No. AAH01631.1
Gene Alias PTMB4, T beta-4, TB4, TB4Xbeta 4, X chromosome, THYB4, thymosin beta 4, X-linked, thymosin beta-4
Host Species Mouse
Immunogen TMSB4X (AAH01631.1, 1 a.a. - 44 a.a.) full-length human protein. MSDKPDMAEIEKFDKSKLKKTETQEKNPLPSKETIEQEKQAGES
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 7114
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.