missing translation for 'onlineSavingsMsg'
Learn More

Thymosin beta 10 Antibody (2E3), Novus Biologicals™

Product Code. 18328418 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328418 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328418 Supplier Novus Biologicals Supplier No. H00009168M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Thymosin beta 10 Monoclonal antibody specifically detects Thymosin beta 10 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thymosin beta 10
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2E3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH16025
Gene Alias MIG12, migration-inducing gene 12, migration-inducing protein 12, PTMB10, TB10, THYB10, thymosin beta 10, thymosin beta-10
Host Species Mouse
Immunogen TMSB10 (AAH16025, 1 a.a. ~ 44 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MADKPDMGEIASFDKAKLKKTETQEKNTLPTKETIEQEKRSEIS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9168
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.