missing translation for 'onlineSavingsMsg'
Learn More

Thymosin alpha 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £255.00

Specifications

Antigen Thymosin alpha 1
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18659936
View Documents
Novus Biologicals
NBP2-99789-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18671797
View Documents
Novus Biologicals
NBP2-99789-50ul
50 μg
£169.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thymosin alpha 1 Polyclonal antibody specifically detects Thymosin alpha 1 in Human samples. It is validated for ELISA
Specifications

Specifications

Thymosin alpha 1
ELISA
Unconjugated
Rabbit
Cell Cycle and Replication
0.2 um filtered solution in PBS
5757
IgG
Antigen and protein A Affinity-purified
ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
Human
alpha (gene sequence 28), gene sequence 28, MGC104802, prothymosin alpha, prothymosin alpha protein, prothymosin, alpha, PTMA prothymosin, alpha, TMSA
Produced in rabbits immunized with purified, recombinant Human Thymosin alpha 1 (Accession#: P06454-1; Ser2-Asp111)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.