missing translation for 'onlineSavingsMsg'
Learn More

Thymolsulfonephthaleindi-(Methyl-Iminodiacetic Acid) Sodium Salt, Pure, Fisher Chemical

Product Code. 10776111 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Jar
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10776111 1 g Glass Jar 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10776111 Supplier Fisher Chemical Supplier No. T/1500/43
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1945-77-3
Color Green
pH 6.2
Physical Form Powder/Solid
Quantity 1 g
Molecular Formula C37H44N2Na4O13S
MDL Number 148905
Synonym methylthymol blue sodium salt, methylthymol blue, n-methylthymol blue, n-methylthymol blue sodium salt, thymolsulfonephthaleindi-methyl-iminodiacetic acid sodium salt, ethyl-3,1-phenylene methylene bis n-carboxymethyl-,s,s-dioxide,tetrasodiu, glycine,n,n inverted exclamation mark-3h-2,1-benzoxathiol-3-ylidenebis 6-hydroxy-2-methyl-5-1-methyl, sodium 2,2',2,2'-1,1-dioxido-3h-benzo c 1,2 oxathiole-3,3-diyl bis 6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene bis methylene bis azanetriyl tetraacetate
InChI Key LGVVZVZPOQJZKT-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]
IUPAC Name 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
Molecular Weight (g/mol) 848.779
PubChem CID 131850582
Formula Weight 844.76g/mol
Packaging Glass Jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.