missing translation for 'onlineSavingsMsg'
Learn More

Thymolphthalein, TRC

Product Code. 30135605
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135605 5 g 5g
30137971 1 g 1g
30003359 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135605 Supplier Toronto Research Chemicals Supplier No. TRCT4130935G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-20-2
Molecular Formula C28 H30 O4
Molecular Weight (g/mol) 430.54
Synonym 3,3-Bis[4-hydroxy-2-methyl-5-(1-methylethyl)phenyl]-1(3H)-isobenzofuranone, 2-[(4-Hydroxy-5-isopropyl-2-methylphenyl)(5-isopropyl-2-methyl-4-oxo-2,5-cyclohexadien-1-ylidene)methyl]benzoic acid, 2',2' '-Diisopropyl-5',5' '-dimethylphenolphthalein, 3,3-Bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one, 3,3-Bis(4-hydroxy-3-isopropyl-6-methyl)phthalide, 3,3-Bis-(4-hydroxy-5-isopropyl-2-methyl-phenyl)-3H-isobenzofuran-1-one
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
SMILES CC(C)c1cc(c(C)cc1O)C2(OC(=O)c3ccccc23)c4cc(C(C)C)c(O)cc4C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C28H30O4/c1-15(2)20-13-23(17(5)11-25(20)29)28(22-10-8-7-9-19(22)27(31)32-28)24-14-21(16(3)4)26(30)12-18(24)6/h7-16,29-30H,1-6H3
Formula Weight 430.2144
Chemical Name or Material Thymolphthalein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.