missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymolphthalein, Pure, Solid, Indicator Grade, Fisher Chemical™
£276.00
Chemical Identifiers
CAS | 125-20-2 |
---|---|
Molecular Formula | C28H30O4 |
Molecular Weight (g/mol) | 430.544 |
MDL Number | 5909 |
InChI Key | LDKDGDIWEUUXSH-UHFFFAOYSA-N |
PubChem CID | 31316 |
IUPAC Name | 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one |
SMILES | CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10766111
|
Fisher Chemical
T/1480/46 |
25 g | |||||||
Description
Chemical Identifiers
125-20-2 | |
430.544 | |
LDKDGDIWEUUXSH-UHFFFAOYSA-N | |
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one |
C28H30O4 | |
5909 | |
31316 | |
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
Specifications
251°C | |
C28H30O4 | |
LDKDGDIWEUUXSH-UHFFFAOYSA-N | |
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one | |
31316 | |
Glass Jar | |
Solid |
125-20-2 | |
5909 | |
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O | |
430.544 | |
430.53g/mol | |
White |