missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymol Blue sodium salt, ACS reagent

Product Code. 10647581 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10647581 25 g Glass bottle 25g
10519292 10 g Glass bottle 10g
10502184 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10647581 Supplier Thermo Scientific Chemicals Supplier No. 420940250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62625-21-2 | C27H29NaO5S | 488.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-21-2
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) 488.57
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
PubChem CID 23692293
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O

Specifications

Melting Point 283.0°C
Clarity Passes Test
Infrared Spectrum Authentic
IUPAC Name sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
Formula Weight 488.58
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval Acid range: from pH 1.2 (pink) to pH 2.8 (yellow) Alkaline range: from pH 8.0 (yellow) to pH 9.2 (blue)
Color Black/Brown or Black/Green to Orange
Quantity 25 g
Physical Form Powder
Chemical Name or Material Thymol Blue sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.