missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymol Blue sodium salt

Product Code. 11426176
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426176 10 g 10g
11436176 50 g 50g
11446176 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426176 Supplier Thermo Scientific Chemicals Supplier No. B23007.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62625-21-2 | C27H29NaO5S | 488.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-21-2
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) 488.57
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
Synonym Thymol Blue, water soluble
PubChem CID 23692293
IUPAC Name sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O

Specifications

Melting Point ∼284°C (decomposition)
Beilstein 3861515
Formula Weight 488.58
Quantity 10 g
Chemical Name or Material Thymol Blue sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.