missing translation for 'onlineSavingsMsg'
Learn More

Thymine, 99%

Product Code. 11916521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11916521 500 g Plastic bottle 500g
10341184 5 g Glass Bottle 5g
10478880 25 g Plastic bottle 25g
10704481 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11916521 Supplier Thermo Scientific Acros Supplier No. 157855000
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65-71-4 | C5H6N2O2 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-71-4
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006026
InChI Key RWQNBRDOKXIBIV-UHFFFAOYSA-N
Synonym thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 1135
ChEBI CHEBI:17821
IUPAC Name 5-methyl-1H-pyrimidine-2,4-dione
SMILES CC1=CNC(=O)NC1=O

Specifications

Melting Point 316.0°C to 317.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 24,353
Merck Index 15,9553
Solubility Information Solubility in water: slightly soluble (4g/L 25°C). Other solubilities: soluble in hot water, readily soluble in alkalies, sparingly soluble in ether, practically insoluble in alcohol
Formula Weight 126.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Thymine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.