missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymidylate Synthase Mouse anti-Human, Clone: rTYMS/1884, Novus Biologicals™
Mouse Monoclonal Antibody
£387.00
Specifications
| Antigen | Thymidylate Synthase |
|---|---|
| Clone | rTYMS/1884 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
Description
Thymidylate Synthase Monoclonal specifically detects Thymidylate Synthase in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Thymidylate Synthase | |
| 0.2 mg/mL | |
| Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing, Transcription Factors and Regulators, Translation Control | |
| 10 mM PBS with 0.05% BSA | |
| 7298 | |
| IgG1 κ | |
| Protein A or G purified |
| rTYMS/1884 | |
| Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.1.1.45, HsT422, MGC88736, thymidylate synthase, thymidylate synthetase, TMSTsase, TSase, TSHST422 | |
| Recombinant human Thymidylate Synthase protein fragment (around aa 60-174) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title