missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thymidylate Synthase Polyclonal specifically detects Thymidylate Synthase in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Thymidylate Synthase |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.1.1.45, HsT422, MGC88736, thymidylate synthase, thymidylate synthetase, TMSTsase, TSase, TSHST422 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Thymidylate Synthase. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?