missing translation for 'onlineSavingsMsg'
Learn More

Thymidine glycol Rabbit anti-Human, Clone: B29, Novus Biologicals™

Product Code. 18302065 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18302065 25 μg 25µg
18363752 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18302065 Supplier Novus Biologicals Supplier No. NBP31104125UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Thymidine glycol Monoclonal specifically detects Thymidine glycol in Human samples. It is validated for ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thymidine glycol
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone B29
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:100
Formulation PBS (pH 7.4), 50% Glycerol
Host Species Rabbit
Immunogen Thymidine glycol polymer
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.