Learn More
Thymidine glycol Antibody (B29), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-11041C
Product Code. 18552107
  • £593.00 / 0.10mL
Description
Thymidine glycol Monoclonal antibody specifically detects Thymidine glycol in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thymidine glycol
Monoclonal
DyLight 650
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Human
Purified
ELISA, Immunohistochemistry
B29
ELISA, Immunohistochemistry
32645-65-1, 5,6-dihydro-5,6-dihydroxythymidine, 5,6-dihydroxy-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,3-diazinane-2,4-dione, 5,6-Dihydroxy-5,6-dihydrothymidine, THD glycol, Thymidine glycol, Thymidine, 5,6-dihydro-5,6-dihydroxy-
Thymidine glycol polymer
0.1 mL
Primary
Store at 4°C in the dark.