missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymidine, 99+%

Product Code. 10053050 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10053050 5 g Glass bottle 5g
10033433 25 g Glass bottle 25g
11986811 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10053050 Supplier Thermo Scientific Chemicals Supplier No. 226740050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-89-5 | C10H14N2O5 | 242.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-89-5
Molecular Formula C10H14N2O5
Molecular Weight (g/mol) 242.23
InChI Key IQFYYKKMVGJFEH-UHFFFAOYNA-N
Synonym thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
PubChem CID 5789
ChEBI CHEBI:17748
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Specifications

Absorbance 0.62 to 0.68 (A250/A260),0.70 to 0.76 (A280/A260)
Melting Point 187.0°C to 189.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 159552
Specific Rotation Condition + 18.50 (20.00°C c=3,H2O)
Specific Rotation + 18.50
Solubility Information Solubility in water: 46g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in methanol, hot ethanol, hot acetone, hot, ethyl acetate and pyridine, sparingly soluble in hot chloroform
Spectrophotometry 98.0 to 102% (lambda max. 267nm,pH 7) molar absorptivity 9600+/-4%,on dry basis
Formula Weight 242.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Thymidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.