missing translation for 'onlineSavingsMsg'
Learn More

Melford Thymidine

Product Code. p-200087227
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 30018831

Brand: Melford T18050100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Store at ambient temperature.

Store at ambient temperature.

Specifications

CAS 50-89-5
Molecular Formula C10H14N2O5
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.