missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THUMPD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588232
This item is not returnable.
View return policy
Description
Immunogen sequence: IVLLLSEDHH RRLTDCKESN IPFNSKDSHT DEPGIKKCLN PEEKTGAFKT ASTSFEASNH KFLDRMSPFG SLVPVECYKV SLGKTDAFIC KYKKSHSSGL; Positive Samples: A-549, HeLa, HT-29, 293T, Mouse brain, Rat liver.
THUMPD2 (THUMP Domain Containing 2) is a Protein Coding gene. Diseases associated with THUMPD2 include Retinitis Pigmentosa 11. Gene Ontology (GO) annotations related to this gene include RNA binding and methyltransferase activity. An important paralog of this gene is THUMPD3.
Specifications
| THUMPD2 | |
| Polyclonal | |
| Unconjugated | |
| Thumpd2 | |
| 2810025A12Rik; C2orf8; SAM-dependent methyltransferase; thum2; THUMP domain containing 2; THUMP domain-containing protein 2; THUMPD2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 313851, 72167, 80745 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.72 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9BTF0, Q9CZB3 | |
| Thumpd2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 404-503 of human THUMPD2 (NP_0795402). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction