missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ THUMPD2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA532045

Product Code. 13436617

  • £446.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

THUMPD2 (THUMP Domain Containing 2) is a Protein Coding gene. Diseases associated with THUMPD2 include Retinitis Pigmentosa 11. Gene Ontology (GO) annotations related to this gene include RNA binding and methyltransferase activity. An important paralog of this gene is THUMPD3.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

THUMPD2
Polyclonal
Unconjugated
Thumpd2
2810025A12Rik; C2orf8; SAM-dependent methyltransferase; thum2; THUMP domain containing 2; THUMP domain-containing protein 2; THUMPD2
Rabbit
Antigen affinity chromatography
RUO
80745
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q9BTF0
Thumpd2
Recombinant protein encompassing a sequence within the center region of human THUMPD2. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.