missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THUMPD1 Antibody (OTI1H6), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74512FR
This item is not returnable.
View return policy
Description
THUMPD1 Monoclonal specifically detects THUMPD1 in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| THUMPD1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| THUMPD1 | |
| Full length recombinant protein of human THUMPD1 (NP_060206) produced in HEK293T cells. | |
| 0.1 mL | |
| 55623.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1H6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DKFZp686C1054, FLJ20274, THUMP domain containing 1, THUMP domain-containing protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction