missing translation for 'onlineSavingsMsg'
Learn More

THUMPD1 Antibody (OTI1H6), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18421568 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421568 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421568 Supplier Novus Biologicals Supplier No. NBP274512AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

THUMPD1 Monoclonal antibody specifically detects THUMPD1 in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen THUMPD1
Applications Western Blot, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone OTI1H6
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias DKFZp686C1054, FLJ20274, THUMP domain containing 1, THUMP domain-containing protein 1
Host Species Mouse
Immunogen Full length recombinant protein of human THUMPD1 (NP_060206) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55623
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.