missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) trifluoromethanesulfonate, 98%

Product Code. 11363868
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363868 2 g 2g
11373868 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363868 Supplier Thermo Scientific Alfa Aesar Supplier No. 040313.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141478-68-4 | C3F9O9S3Tm | 616.124 g/mol

A raw material used in synthesis of chemicals .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141478-68-4
Molecular Formula C3F9O9S3Tm
Molecular Weight (g/mol) 616.124
MDL Number MFCD00209617
InChI Key PBASUZORNBYVFM-UHFFFAOYSA-K
Synonym thulium iii trifluoromethanesulfonate, thulium iii triflate, trifluoromethanesulfonic acid thulium iii salt, thulium triflate, tm otf 3, acmc-1c8m6, thulium 3+ tritriflate, thulium 3+ ; trifluoromethanesulfonate, thulium trifluoromethanesulfonate, thulium iii tris trifluoromethanesulfonate
PubChem CID 2733936
IUPAC Name thulium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tm+3]

Specifications

Physical Form Powder
Quantity 2 g
Linear Formula Tm(OSO2CF3)3
Solubility Information Soluble in water.
Formula Weight 616.13
Percent Purity 98%
Chemical Name or Material Thulium(III) trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.