missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III)-sulfate octahydrate, 99.99%, (trace metal basis), Thermo Scientific™

Product Code. 10184160 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10184160

Brand: Thermo Scientific Acros 194290050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

CAS 13778-40-0
Color Green
Physical Form Fine Crystalline Powder
Quantity 5 g
Assay Percent Range 99.99%
Molecular Formula O12S3Tm2·8H2O
Linear Formula Tm2(SO4)3·8H2O
MDL Number MFCD00149881
Merck Index 15, 9548
Synonym thulium sulfate octahydrate, unii-8zgz55l20j, thulium sulfate octahydrate mi, acmc-20aji8, thulium sulfate tm2 so4 3 octahydrate, thulium sulfate hydrate tm2 so4 3.8h2o, sulfuric acid, thulium 3+ salt 3:2 , octahydrate, thulium sulfate-water 2/3/8, thulium sulfate,, sulfuric acid thulium iii salt octahydrate
InChI Key GPBYLGMJMIXKMU-UHFFFAOYSA-H
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Tm+3].[Tm+3]
IUPAC Name thulium(3+);trisulfate;octahydrate
Molecular Weight (g/mol) 770.17
PubChem CID 71310074
Formula Weight 770.17
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Thulium(III)-sulfate octahydrate, 99.99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.