missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) fluoride, ultra dry, 99.99% (REO), Thermo Scientific™

Product Code. 11348937
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348937 5 g 5g
11358937 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348937 Supplier Thermo Scientific Alfa Aesar Supplier No. 038615.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-79-7
Molecular Formula F3Tm
Molecular Weight (g/mol) 225.93
MDL Number MFCD00049611
InChI Key AATUHDXSJTXIHB-UHFFFAOYSA-K
IUPAC Name thulium(3+) trifluoride
SMILES [F-].[F-].[F-].[Tm+3]

Specifications

Boiling Point 2200°C
Melting Point 1158°C
Physical Form Powder
Quantity 5 g
UN Number UN3288
Solubility Information It is insoluble in water.
Formula Weight 225.93
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 7.9 g/mL
Assay (REO)
Chemical Name or Material Thulium(III) fluoride, Ultra dry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.