missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) fluoride, anhydrous, 99.9% (REO)

Product Code. 11396898
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396898 1 g 1g
11306908 5 g 5g
11316908 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396898 Supplier Thermo Scientific Alfa Aesar Supplier No. 013652.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13760-79-7 | F3Tm | 225.93 g/mol

Thulium(III) fluoride, anhydrous is used for glass doping and to enhance mid-IR emission in oxyfluoride glass ceramics. It is also used to improve ultraviolet and visible upconversion fluorescence in fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-79-7
Molecular Formula F3Tm
Molecular Weight (g/mol) 225.93
MDL Number MFCD00049611
InChI Key AATUHDXSJTXIHB-UHFFFAOYSA-K
IUPAC Name thulium(3+) trifluoride
SMILES [F-].[F-].[F-].[Tm+3]

Specifications

Boiling Point 2200°C
Melting Point 1158°C
Physical Form Crystalline
Quantity 1 g
UN Number UN3288
Solubility Information Insoluble in water
Formula Weight 225.93
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.9 g/mL
Assay (REO)
Chemical Name or Material Thulium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.