missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) chloride hydrate, REacton™, 99.9% (REO)

Product Code. p-4904551
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326238 2 g 2g
11336238 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326238 Supplier Thermo Scientific Alfa Aesar Supplier No. 011200.04
Add to basket
This item is not returnable. View return policy

CAS: 19423-86-0 | Cl3Tm | 275.28 g/mol

Thulium(III) chloride hydrate is used in proteomics research. It is also used in the production of glass, phosphors and lasers. Further, it serves as an important dopant for fiber amplifiers. Further, it is also used to prepare thulium(III) oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-86-0
Molecular Formula Cl3Tm
Molecular Weight (g/mol) 275.28
MDL Number MFCD00011283
InChI Key ILOTUXNTERMOJL-UHFFFAOYSA-K
IUPAC Name thulium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Tm+3]

Specifications

Color Green
Physical Form Crystalline Aggregates
Quantity 2 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water and alcohol.
Formula Weight 275.29 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Thulium(III) chloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.