missing translation for 'onlineSavingsMsg'
Learn More

Thulium foil, 0.1mm (0.004 in.) thick, 99.9% (REO), Thermo Scientific™

Product Code. 11355779
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-30-4
Molecular Formula Tm
Molecular Weight (g/mol) 168.93
MDL Number MFCD00011281
InChI Key FRNOGLGSGLTDKL-UHFFFAOYSA-N
Synonym tulio, unii-8rkc5ati4p, 8rkc5ati4p, atom, pieces, powder, foil, acmc-20ajig, thulium, ingot, pieces, sublimed dendritic
PubChem CID 23961
ChEBI CHEBI:33380
IUPAC Name thulium
SMILES [Tm]

Specifications

Color Silver to Gray
UN Number UN3178
Quantity 50 x 50 mm
Percent Purity 99.9%
Odor Odorless
Weight ≈0.58g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Thulium foil, 0.1mm (0.004 in.) thick
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'