Learn More
THTPA (Human) Matched Antibody Pair, Abnova™
Matched Antibody Pair
Brand: Abnova H00079178-AP11.1Set
This item is not returnable.
View return policy
Specifications
| THTPA | |
| Unconjugated | |
| MGC2652/THTP/THTPASE | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-THTPA (100 ug) 2. Detection antibody: mouse monoclonal anti-THTPA, IgG1 Kappa (20 ug) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| thiamine triphosphatase | |
| THTPA | |
| RUO | |
| 79178 | |
| Human | |
| Ab Pair |