missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THSD7A Monoclonal Antibody (CL3778)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531390
This item is not returnable.
View return policy
Description
Immunogen sequence: VCQSSPCEAE ELRYSLHVGP WSTCSMPHSR QVRQARRRGK NKEREKDRSK GVKDPEAREL IKKKRNRNRQ NRQENKYWDI QIGYQTREVM CINKTGKAAD LSFCQQEKLP MTFQSC Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 83%.
The protein encoded by this gene is found almost exclusively in endothelial cells from placenta and umbilical cord. The encoded protein appears to interact with alpha(V)beta(3) integrin and paxillin to inhibit endothelial cell migration and tube formation. This protein may be involved in cytoskeletal organization. Variations in this gene may be associated with low bone mineral density in osteoporosis. [provided by RefSeq, Aug 2010].
Specifications
| THSD7A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UPZ6 | |
| THSD7A | |
| Recombinant protein fragment of Human THSD7A (Product #RP-93709) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| CL3778 | |
| Unconjugated | |
| THSD7A | |
| KIAA0960; thrombospondin type 1 domain containing 7A; thrombospondin type-1 domain-containing protein 7A; Thrombospondin type-1 domain-containing protein 7A, soluble form; thrombospondin, type I, domain containing 7A; THSD7A | |
| Mouse | |
| Protein A | |
| RUO | |
| 221981 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction