Learn More
THSD7A Antibody (CL3778) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61638CL1
Product Code. 30303100
  • £887.00 / 0.10mL
Description
THSD7A Monoclonal antibody specifically detects THSD7A in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
THSD7A
Monoclonal
CoraFluor 1
thrombospondin, type I, domain containing 7A
This antibody was developed using a recombinant protein derived from Q9UPZ6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology
221981
Store at 4⪚C in the dark. Do not freeze.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL3778
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified