missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Thrombospondin-1 Antibody (A65M), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05185AF350
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Thrombospondin-1 Monoclonal antibody specifically detects Thrombospondin-1 in Human, Bovine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Tekniske data
| Thrombospondin-1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| THBS1 | |
| Extracellular matrix material from cultured bovine corneal endothelial cells. | |
| 0.1 mL | |
| Primary | |
| Highly specific for the low-calcium (0.1 mmol/L) conformation of human TSP1. There is no evidence of cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin). | |
| Store at 4°C in the dark. |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| A65M | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| THBS, THBS-1, thrombospondin 1, thrombospondin-1p180, TSP1thrombospondin-1, TSPTSP-1 | |
| Mouse | |
| Protein A or G purified | |
| Cancer | |
| 7057 | |
| Human, Bovine | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion