missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THSD1 Antibody (541213), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5178PEATT594
This item is not returnable.
View return policy
Description
THSD1 Monoclonal antibody specifically detects THSD1 in Human samples. It is validated for Flow Cytometry
Specifications
| THSD1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 541213 | |
| Flow Cytometry | |
| thrombospondin type-1 domain-containing protein 1, thrombospondin, type I, domain 1, thrombospondin, type I, domain containing 1,4833423O18Rik, TMTSPMGC74971, Transmembrane molecule with thrombospondin module, UNQ3010 | |
| Mouse myeloma cell line NS0-derived recombinant human THSD1 isoform 2, Glu25-Ile361, Accession # NP_954872 | |
| 0.1 mL | |
| Cancer | |
| 55901 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction