missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THSD1 Antibody (122), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90566F
This item is not returnable.
View return policy
Description
THSD1 Monoclonal specifically detects THSD1 in Mouse samples. It is validated for ELISA.
Specifications
| THSD1 | |
| Monoclonal | |
| FITC | |
| thrombospondin type-1 domain-containing protein 1, thrombospondin, type I, domain 1, thrombospondin, type I, domain containing 1,4833423O18Rik, TMTSPMGC74971, Transmembrane molecule with thrombospondin module, UNQ3010 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse THSD1 (NP_062522.1; Met 1-Asn 412). | |
| 0.1 mL | |
| Cancer | |
| 55901 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 122 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction