missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thromboxane A2 R/TBXA2R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31106
This item is not returnable.
View return policy
Description
Thromboxane A2 R/TBXA2R Polyclonal specifically detects Thromboxane A2 R/TBXA2R in Human samples. It is validated for Western Blot.
Specifications
| Thromboxane A2 R/TBXA2R | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| Prostanoid TP receptor, thromboxane A2 receptor, TXA2-R | |
| Rabbit | |
| 37 kDa | |
| 0.1 mL | |
| GPCR | |
| 6915 | |
| Human | |
| IgG |
| Western Blot | |
| 0.55mg/mL | |
| Western Blot 1:500-1:3000 | |
| P21731 | |
| TBXA2R | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human TXA2R. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction