missing translation for 'onlineSavingsMsg'
Learn More

Thrombospondin-4 Antibody, Novus Biologicals™

Product Code. 18358347 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358347 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358347 Supplier Novus Biologicals Supplier No. H00007060B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Thrombospondin-4 Polyclonal antibody specifically detects Thrombospondin-4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin-4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_003239.2
Gene Alias thrombospondin 4, thrombospondin-4, TSP4
Host Species Mouse
Immunogen THBS4 (NP_003239.2, 1 a.a. - 961 a.a.) full-length human protein. MLAPRGAAVLLLHLVLQRWLAAGAQATPQVFDLLPSSSQRLNPGALLPVLTDPALNDLYVISTFKLQTKSSATIFGLYSSTDNSKYFEFTVMGRLNKAILRYLKNDGKVHLVVFNNLQLADGRRHRILLRLSNLQRGAGSLELYLDCIQVDSVHNLPRAFAGPSQKPETIELRTFQRKPQDFLEELKLVVRGSLFQVASLQDCFLQQSEPLAATGTGDFNRQFLGQMTQLNQLLGEVKDLLRQQVKETSFLRNTIAECQACGPLKFQSPTPSTVVPPAPPAPPTRPPRRCDSNPCFRGVQCTDSRDGFQCGPCPEGYTGNGITCIDVDECKYHPCYPGVHCINLSPGFRCDACPVGFTGPMVQGVGISFAKSNKQVCTDIDECRNGACVPNSICVNTLGSYRCGPCKPGYTGDQIRGCKAERNCRNPELNPCSVNAQCIEERQGDVTCVCGVGWAGDGYICGKDVDIDSYPDEELPCSARNCKKDNCKYVPNSGQEDADRDGIGDACDEDADGDGILNEQDNCVLIHNVDQRNSDKDIFGDACDNCLSVLNNDQKDTDGDGRGDACDDDMDGDGIKNILDNCPKFPNRDQRDKDGDGVGDACDSCPDVSNPNQSDVDNDLVGDSCDTNQDSDGDGHQDSTDNCPTVINSAQLDTDKDGIGDECDDDDDNDGIPDLVPPGPDNCRLVPNPAQEDSNSDGVGDICESDFDQDQVIDRIDVCPENAEVTLTDFRAYQTVVLDPEGDAQIDPNWVVLNQGMEIVQTMNSDPGLAVGYTAFNGVDFEGTFHVNTQTDDDYAGFIFGYQDSSSFYVVMWKQTEQTYWQATPFRAVAEPGIQLKAVKSKTGPGEHLRNSLWHTGDTSDQVRLLWKDSRNVGWKDKVSYRWFLQHRPQVGYIRVRFYEGSELVADSGVTIDTTMRGGRLGVFCFSQENIIWSNLKYRCNDTIPEDFQEFQTQNFDRFDN
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7060
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.