Learn More
Thrombospondin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-68953
 View more versions of this product
Product Code. 18661938
  • £513.00 / 100µL
Description
Thrombospondin-3 Polyclonal antibody specifically detects Thrombospondin-3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thrombospondin-3
Polyclonal
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
MGC119565, thrombospondin 3, thrombospondin-3, TSP3MGC119564
This antibody was developed against a recombinant protein corresponding to amino acids: VGALSECPFQGDESIHSAVTNALHSILGEQTKALVTQLTLFNQILVELRDDIRDQVKEMSLIRNTIMECQVCGFHEQRS
100 μg
Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction
7059
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS (pH 7.2) and 40% Glycerol
Rabbit
Protein A purified
RUO
Primary
Human
Purified