Learn More
Invitrogen™ Thrombospondin 1 Monoclonal Antibody (A6.1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA513398
Description
MA5-13398 targets Thrombospondin in FACS, ICC/IF, IHC (P), IM, IP, and WB applications and shows reactivity with Bovine, Canine, Equine, Human, mouse, Ovine, Porcine, and Rat samples. The MA5-13398 immunogen is reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets.
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
Specifications
| Thrombospondin 1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P07996, P35441, Q28178 | |
| THBS1 | |
| Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets. | |
| 200 μL | |
| Primary | |
| Bovine, Canine, Horse, Human, Mouse, Sheep, Pig, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| A6.1 | |
| Unconjugated | |
| THBS1 | |
| glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100057478, 101106433, 21825, 281530, 445442, 487486, 492313, 7057 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.