missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombospondin 1 Monoclonal Antibody (A4.1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513377
This item is not returnable.
View return policy
Description
MA5-13377 targets Thrombospondin in FACS, IF, and WB applications and shows reactivity with Bovine, Chicken, Human, mouse, and Rat samples. The MA5-13377 immunogen is calcium-replete, native purified human TSP from the supernatant of thrombin-activated platelets.
Thrombospondin 1 Monoclonal antibody specifically detects Thrombospondin 1 in Bovine, Chicken, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.
Specifications
| Thrombospondin 1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P07996, P35441, Q28178 | |
| THBS1 | |
| Calcium-replete, native purified human TSP from the supernatant of thrombin-activated platelets | |
| 500 μL | |
| Primary | |
| Bovine, Chicken, Human, Mouse, Rat | |
| Antibody | |
| IgM |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| A4.1 | |
| Unconjugated | |
| THBS1 | |
| glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 21825, 281530, 373987, 445442, 7057 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction