missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombospondin-1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-52410
This item is not returnable.
View return policy
Description
Thrombospondin-1 Polyclonal antibody specifically detects Thrombospondin-1 in Human, Mouse, Porcine, Bovine, Equine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Peptide ELISA
Specifications
| Thrombospondin-1 | |
| Polyclonal | |
| Western Blot 0.3-1 μg/mL, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Paraffin 3.75 μg/mL, Peptide ELISA 1:128000 | |
| P07996 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Bovine, Equine | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| TBS (pH 7.3) and 0.5% BSA | |
| THBS, THBS-1, thrombospondin 1, thrombospondin-1p180, TSP1thrombospondin-1, TSPTSP-1 | |
| Peptide with sequence NRIPESGGDNSVFD-C, from the N Terminus of the Thrombospondin-1 protein sequence according to NP_003237.2. | |
| 0.05 mg | |
| Cancer | |
| 7057 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction