missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombopoietin/THPO Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33515-100ul
This item is not returnable.
View return policy
Description
Thrombopoietin/THPO Monoclonal antibody specifically detects Thrombopoietin/THPO in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Thrombopoietin/THPO | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDFC-mpl ligand, ML, MPL ligand, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, thrombopoietin, thrombopoietin nirs variant 1, TPOMKCSF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-353 of mouse Thrombopoietin/THPO (NP_033405.1).,, Sequence:, SPVAPACDPRLLNKLLRDSHLLHSRLSQCPDVDPLSIPVLLPAVDFSLGEWKTQTEQSKAQDILGAVSLLLEGVMAARGQLEPSCLSSLLGQLSGQVRLLLGALQGLLGTQLPLQGRTTAHKDPNALFLSLQQLLRGKVRFLLLVEGPTLCVRRTLPTTAVPSSTSQLLTLNKFPNRTSGLLETNFSVTARTAGPGLLSRLQGFRVKITPGQLNQTSRSPVQISGYLNRTHGPVNGTHGLFAGTSLQTLEASDISPGAFNKGSLAFNLQGGLPPSPSLAPDGHTPFPPSPALPTTHGSPPQLHPLFPDPSTTMPNSTAPHPVTMYPHPRNLS | |
| 100 μL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7066 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction