missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombopoietin/THPO Antibody (MM0566-1Z3), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11922DL594
This item is not returnable.
View return policy
Description
Thrombopoietin/THPO Monoclonal antibody specifically detects Thrombopoietin/THPO in Human samples. It is validated for Western Blot
Specifications
| Thrombopoietin/THPO | |
| Monoclonal | |
| DyLight 594 | |
| Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDFC-mpl ligand, ML, MPL ligand, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, thrombopoietin, thrombopoietin nirs variant 1, TPOMKCSF | |
| Human recombinant Thrombopoietin/THPO | |
| 0.1 mL | |
| Biologically Active Proteins, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 7066 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| MM0566-1Z3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction