missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombopoietin/THPO Antibody (444), Alexa Fluor™ 647, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Thrombopoietin/THPO Monoclonal specifically detects Thrombopoietin/THPO in Mouse samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Thrombopoietin/THPO |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 444 |
| Conjugate | Alexa Fluor 647 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDFC-mpl ligand, ML, MPL ligand, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, thrombopoietin, thrombopoietin nirs variant 1, TPOMKCSF |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Thrombopoietin/THPO (NP_033405.1; Met 1-Thr 356). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?