missing translation for 'onlineSavingsMsg'
Learn More

Thrombopoietin/THPO Antibody (34817) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30732801 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732801 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732801 Supplier Novus Biologicals Supplier No. FAB288I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Thrombopoietin/THPO Monoclonal antibody specifically detects Thrombopoietin/THPO in Human samples. It is validated for ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombopoietin/THPO
Applications ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 34817
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDFC-mpl ligand, ML, MPL ligand, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, thrombopoietin, thrombopoietin nirs variant 1, TPOMKCSF
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Thrombopoietin/Tpo, Ser22-Gly353, Accession # P40225
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7066
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.