missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombopoietin R/Tpo R Rabbit anti-Human, Mouse, Rat, Clone: 5Q9Z6, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | Thrombopoietin R/Tpo R |
|---|---|
| Clone | 5Q9Z6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18333626
|
Novus Biologicals
NBP3-15880-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18387672
|
Bio-Techne
NBP3-15880-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thrombopoietin R/Tpo R Monoclonal antibody specifically detects Thrombopoietin R/Tpo R in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Thrombopoietin R/Tpo R | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| CD Markers, Immunology, Stem Cells | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4352 | |
| IgG | |
| Affinity purified |
| 5Q9Z6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CD110, CD110 antigen, C-MPL, Myeloproliferative leukemia protein, myeloproliferative leukemia virus oncogene, Proto-oncogene c-Mpl, thrombopoietin receptor, TPO-R, TPORMPLV | |
| A synthetic peptide corresponding to a sequence within amino acids 536-635 of human Thrombopoietin R/Tpo R (P40238). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title