missing translation for 'onlineSavingsMsg'
Learn More

Thrombopoietin R/Tpo R Antibody (1H2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-37529

 View more versions of this product

Product Code. 18135448

  • £440.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Thrombopoietin R/Tpo R Monoclonal specifically detects Thrombopoietin R/Tpo R in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thrombopoietin R/Tpo R
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready
CD110, CD110 antigen, C-MPL, Myeloproliferative leukemia protein, myeloproliferative leukemia virus oncogene, Proto-oncogene c-Mpl, thrombopoietin receptor, TPO-R, TPORMPLV
Mouse
71.2 kDa
0.1 mL
Primary
Human
IgG2b
Western Blot, Flow Cytometry, ELISA, CyTOF
1H2
Unconjugated
PBS with 0.05% Sodium Azide
MPL
Purified recombinant fragment of human MPL expressed in E. Coli.
Affinity Purified
RUO
4352
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only