missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Thrombopoietin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116038
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total THPO.
Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
Specifications
| Thrombopoietin | |
| Polyclonal | |
| Unconjugated | |
| THPO | |
| C-mpl ligand; Megakaryocyte colony-stimulating factor; megakaryocyte growth and development factor; megakaryocyte stimulating factor; Mgdf; MKCSF; ML; MPL ligand; Mpllg; MSA; Myeloproliferative leukemia virus oncogene ligand; prepro-thrombopoietin; TDH2A; THCYT1; THPO; Thrombopoietin; thyroid microsomal antigen; thyroid peroxidase; thyroperoxidase; Tpo; TPX | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 108348162, 21832, 7066, 81811 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P40225, P40226, P49745 | |
| THPO | |
| A synthesized peptide derived from human THPO(Accession P40225), corresponding to amino acid residues T257-F307. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction