missing translation for 'onlineSavingsMsg'
Learn More

Thrombomodulin / CD141 Mouse, Clone: THBD/1591, enQuireBio™

Mouse Monoclonal Antibody

£181.83 - £423.29

Specifications

Antigen Thrombomodulin / CD141
Clone THBD/1591
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15900968
View Documents
EnquireBio
7056-MSM1-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15910968
View Documents
EnquireBio
7056-MSM1-P1
100 μg
£423.29
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Thrombomodulin / CD141
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
7056
Recombinant human Thrombomodulin protein fragment (aa69-194)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
100kDa
THBD/1591
Immunohistochemistry
Unconjugated
RUO
NP_000352
THBD
IgG2b κ
Protein A/G Purified
Cell Surface and Cytoplasmic
THP1 Cells. Placenta, Angiosarcoma, Bladder or Cervical Carcinoma
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.